4-Amino-L-phenylalanine HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phe(4-NH₂)-OH, H-p-Amino-Phe-OH, H-Phe(4-NH2)-OH

4001637.0005 4001637.0025
F-1225.0005BAEA 261.28 CAD
F-1225.0005BA F-1225.0025BA
4-Amino-L-phenylalanine HCl
4-Amino-L-phenylalanine

Sequence: H-p-Amino-Phe-OH · HCl


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Aph derivatives.

Formula: C₉H₁₂N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00069927
CAS Number: 943-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR