Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Supplier: BACHEM AMERICAS INC

4006587.0050 4006587.1000
F-1215.0050BAEA 570.65 CAD
F-1215.0050BA F-1215.1000BA
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Proteins and Peptides

Sequence: [(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine.


  • EINECS: 261-529-2


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655). Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


Synonym(s): Bestatin#Ubenimex

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR