Lysinoalanine · 2 HCl (diastereomeric mixture: LL + LD)

Supplier: BACHEM AMERICAS INC

4008365.0050 4008365.0250
F-1195.0050BAEA 593.81 CAD
F-1195.0050BA F-1195.0250BA
Lysinoalanine · 2 HCl (diastereomeric mixture: LL + LD)
Lysinoalanine · 2 HCl (diastereomeric mixture: LL + LD)

Sequence: H-Lys(DL-2-amino-2-carboxyethyl)-OH · 2 HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Synonym(s): LAL · 2 HCl

Formula: C₉H₁₉N₃O₄·2HCl
MW: 306.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058086
CAS Number: 4418-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR