(S)-(+)-2-Aminobutyric acid

Supplier: BACHEM AMERICAS INC

Synonyms: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

4030313.0005 4030313.0025
F-1190.0005BAEA 206.51 CAD
F-1190.0005BA F-1190.0025BA
(S)-(+)-2-Aminobutyric acid
(S)-(+)-2-Aminobutyric acid

Sequence: H-Abu-OH

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS Number: 1492-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR