DL-α-Aminoadipic acid

Supplier: BACHEM AMERICAS INC

4001419.0005 4001419.0025
F-1185.0005BAEA 271.52 CAD
F-1185.0005BA F-1185.0025BA
DL-α-Aminoadipic acid
DL-α-Aminoadipic acid

Sequence: H-DL-Aad-OH
Synonym(s): DL-α-Aminoadipic acid#(RS)-2-Aminohexanedioic acid#H-DL-Homoglu-OH

Formula: C₆H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00063119
CAS Number: 542-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR