D-allo-Threonine

Supplier: BACHEM AMERICAS INC

4008353.0250 4008353.1000
F-1180.0250BAEA 387.12 CAD
F-1180.0250BA F-1180.1000BA
D-allo-Threonine
D-allo-Threonine

Sequence: H-D-allo-Thr-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.


D-aThr is a constituent of the marine cytotoxic depsipeptide kahalalide F, which showed a remarkable antitumoral activity.

Formula: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00004526
CAS Number: 24830-94-2
EINECS: 246-488-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR