L-allo-Threonine

Supplier: BACHEM AMERICAS INC

Synonyms: H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH

4008379.0250 4008379.1000
F-1175.0250BAEA 206.51 CAD
F-1175.0250BA F-1175.1000BA
L-allo-Threonine
L-allo-Threonine

Sequence: H-allo-Thr-OH


Bachem also offers phosphothreonine derivatives.
aThr derivatives.

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064268
CAS Number: 28954-12-3
EINECS: 249-327-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR