L(+)-allo-Isoleucine

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,3R)-(+)-allo-Isoleucine, L-Alloisoleucine, (2S,3R)-2-Amino-3-methylpentanoic acid

4006513.0250 4006513.1000 4006513.5000
F-1160.0250BAEA 206.51 CAD
F-1160.0250BA F-1160.1000BA F-1160.5000BA
L(+)-allo-Isoleucine
L(+)-allo-Isoleucine

Sequence: H-allo-Ile-OH

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
Storage Temperature: Ambient
MDL Number: MFCD00066446
CAS Number: 1509-34-8
EINECS: 216-142-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR