Methyl-3-aminopropanoate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: beta-Alanine methyl ester hydrochloride, H-β-Ala-OMe·HCl, β-Alanine methyl ester hydrochloride, 3-Aminopropionic acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, Methyl 3-aminopropanoate hydrochloride, Methyl 3-Aminopropionate Hydrochloride, H-β-Ala-OMe.HCl

4002283.0025 4002283.0100
F-1155.0025BAEA 232.35 CAD
F-1155.0025BA F-1155.0100BA
Methyl-3-aminopropanoate hydrochloride
Methyl-3-aminopropanoate hydrochloride

Sequence: H-β-Ala-OMe · HCl


Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).

Formula: C₄H₉NO₂·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00039060
CAS Number: 3196-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR