Ethyl-3-aminopropanoate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: Ethyl 3-aminopropanoate hydrochloridebeta-Alanine ethyl ester hydrochlorideEthyl 3-aminopropionate HCl3-Aminopropionic Acid Ethyl Ester HydrochlorideEthyl 3-Aminopropionate HydrochlorideH-β-Ala-OEt·HCl

4001806.0025
F-1140.0025BAEA 198.38 CAD
F-1140.0025BA
Ethyl-3-aminopropanoate hydrochloride
Ethyl-3-aminopropanoate hydrochloride

Sequence: H-β-Ala-OEt · HCl


Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).

Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012909
CAS Number: 4244-84-2
EINECS: 224-203-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR