β-Alanine-tert-butylester hydrochloride

Supplier: BACHEM AMERICAS INC

4010013.0005 4010013.0025
F-1135.0005BAEA 211.67 CAD
F-1135.0005BA F-1135.0025BA
β-Alanine-tert-butylester hydrochloride
beta-Alanine-tert-butylester hydrochloride

Sequence: H-β-Ala-OtBu · HCl


Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).

Formula: C₇H₁₆ClNO₂
Storage Temperature: Freezer
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR