D-Alanine

Supplier: BACHEM AMERICAS INC

4029158.0025 4029158.0100 4029158.0250
F-1100.0025BAEA 377.3 CAD
F-1100.0025BA F-1100.0100BA F-1100.0250BA
D-Alanine
D-Alanine

Sequence: H-D-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₃H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00008077
CAS Number: 338-69-2
EINECS: 206-418-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR