N-Acetyl-L-hydroxyproline

Supplier: BACHEM AMERICAS INC

4012417.0025 4012417.0100
F-1040.0025BAEA 926.76 CAD
F-1040.0025BA F-1040.0100BA
N-Acetyl-L-hydroxyproline
N-Acetyl-L-hydroxyproline

Sequence: Ac-Hyp-OH


Hyp denotes L-trans-hydroxyproline.


Synonym(s): Oxaceprol

Formula: C₇H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037339
CAS Number: 33996-33-7
EINECS: 251-780-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR