cis-4-Hydroxy-L-(-)-proline

Supplier: BACHEM AMERICAS INC

Synonyms: L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline

4011503.0001 4011503.0005
F-1025.0001BAEA 529.57 CAD
F-1025.0001BA F-1025.0005BA
cis-4-Hydroxy-L-(-)-proline
cis-4-Hydroxy-L(-)-proline

Sequence: H-cis-Hyp-OH


Hyp denotes L-trans-hydroxyproline.


Synonym(s): (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 257
Storage Temperature: Ambient
MDL Number: MFCD00064319
CAS Number: 618-27-9
EINECS: 210-542-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR