N-Acetyl-D-alanine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Ac-D-Ala-OH, Ac-D-Ala-OH

4002334.0001 4002334.0005 4002334.0025
F-1000.0001BAEA 180.91 CAD
F-1000.0001BA F-1000.0005BA F-1000.0025BA
N-Acetyl-D-alanine
N-Acetyl-D-alanine

Sequence: Ac-D-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₅H₉NO₃
MW: 131.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063133
CAS Number: 19436-52-3
EINECS: 243-066-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR