Targinine

Supplier: BACHEM AMERICAS INC

4057243.0050 4057243.0250
E-3745.0050BAEA 439.41 CAD
E-3745.0050BA E-3745.0250BA
Targinine
Targinine

Sequence: H-Arg(Me)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also E-2770, H-Arg(Me)-OH · HCl, in the subfamily 'Arginine Substrates and Inhibitors'.


Synonym(s): L-NMMA#Tilarginine#Targinine

Formula: C₇H₁₆N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD00069311
CAS Number: 17035-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR