N-Acetyl-L-asparagine

Supplier: BACHEM AMERICAS INC

4042263.0005 4042263.0025
E-3705.0005BAEA 754.97 CAD
E-3705.0005BA E-3705.0025BA
N-Acetyl-L-asparagine
N-Acetyl-L-asparagine

Sequence: Ac-Asn-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".


Substrate for L-isoaspartyl/ D-aspartyl methyltransferase (E.C. 2.1.1.77).

Formula: C₆H₁₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00066023
CAS Number: 4033-40-3
EINECS: 223-716-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR