N-(p-Toluenesulfonyl)glycine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

4000075.0025 4000075.0100
E-3700.0025BAEA 387.21 CAD
E-3700.0025BA E-3700.0100BA
N-(p-Toluenesulfonyl)glycine
N-(p-Toluenesulfonyl)glycine

Sequence: Tos-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


The tosyl group is a suitable N-protection group in the Minisci radical alkylation.

Formula: C₉H₁₁NO₄S
MW: 229.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045898
CAS Number: 1080-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR