(S)-Methyl-2-(methylamino)propanoate hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: N-Me-Ala-OMe

4041784.0001 4041784.0005
E-3685.0001BAEA 232.35 CAD
E-3685.0001BA E-3685.0005BA
(S)-Methyl-2-(methylamino)propanoate hydrochloride
(S)-Methyl-2-(methylamino)propanoate hydrochloride

Sequence: N-Me-Ala-OMe


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₅H₁₂ClNO₂
MW: 153.606 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04973252
CAS Number: 35023-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR