Ac-Lys(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4042560.0001 4042560.0005
E-3680.0001BAEA 516.34 CAD
E-3680.0001BA E-3680.0005BA
Ac-Lys(Fmoc)-OH
Ac-Lys(Fmoc)-OH

Sequence: Ac-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₃H₂₆N₂O₅
MW: 410.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458546
CAS Number: 148101-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR