(2S)-2-Amino-3-[4-(hydroxysulfonyloxy)phenyl]propanoic acid

Supplier: BACHEM AMERICAS INC

4033889.0001 4033889.0005
E-3645.0001BAEA 948.9 CAD
E-3645.0001BA E-3645.0005BA
(2S)-2-Amino-3-[4-(hydroxysulfonyloxy)phenyl]propanoic acid
(2S)-2-Amino-3-[4-(hydroxysulfonyloxy)phenyl]propanoic acid

Sequence: H-Tyr(SO₃H)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Bachem also offers a protected stable sulfotyrosine mimic, Fmoc-4-sulfomethyl-Phe(Tce)-OH (B-4110).


Sulfotyrosine has been used by Liu et al. for conveniently obtaining tyrosine-sulfated proteins by expression in E. coli with an expanded genetic code.

Formula: C₉H₁₁NO₆S
Storage Temperature: Freezer
MDL Number: MFCD00191024
CAS Number: 146386-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR