H-His(1-Trt)-OtBu

Supplier: BACHEM AMERICAS INC

4033044.0001 4033044.0005
E-3600.0001BAEA 542.13 CAD
E-3600.0001BA E-3600.0005BA
H-His(1-Trt)-OtBu
H-His(1-Trt)-OtBu

Sequence: H-His(1-Trt)-OtBu


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): H-His(τ-Trt)-OtBu

Formula: C₂₉H₃₁N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD03093489
CAS Number: 304698-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR