Phosphothreonine

Supplier: BACHEM AMERICAS INC

4029206.0001 4029206.0005
E-3575.0001BAEA 387.21 CAD
E-3575.0001BA E-3575.0005BA
Phosphothreonine
Phosphothreonine

Sequence: H-Thr(PO₃H₂)-OH


Bachem also offers phosphothreonine derivatives.
Phosphothreonine is a potential N-terminal site for chemical ligation.

Formula: C₄H₁₀NO₆P
Storage Temperature: Freezer
MDL Number: MFCD00069578
CAS Number: 1114-81-4
EINECS: 214-217-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR