L-Glutamic acid g-methyl ester a-amide hydrochloride

Supplier: BACHEM AMERICAS INC

4028874.0001 4028874.0005
E-3540.0001BAEA 180.91 CAD
E-3540.0001BA E-3540.0005BA
L-Glutamic acid g-methyl ester a-amide hydrochloride
L-Glutamic acid g-methyl ester a-amide hydrochloride

Sequence: H-Glu(OMe)-NH₂ · HCl


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): H-Isogln-OMe · HCl#L-Isoglutamine-methyl ester · HCl

Formula: C₆H₁₃ClN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD02094379
CAS Number: 70830-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR