Aloc-Ala-OH·DCHA

Supplier: BACHEM AMERICAS INC

4028865.0001 4028865.0005
E-3530.0001BAEA 439.41 CAD
E-3530.0001BA E-3530.0005BA
Aloc-Ala-OH·DCHA
Aloc-Ala-OH·DCHA

Sequence: Aloc-Ala-OH · DCHA


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₇H₁₁NO₄·C₁₂H₂₃N
Storage Temperature: Freezer
MDL Number: MFCD02259515
CAS Number: 90508-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR