H-Glu(OME)-OtBu·HCl
Supplier: BACHEM AMERICAS INC
4000148.0001
4000148.0005
E-3515.0001BAEA
284.04
CAD
E-3515.0001BA
E-3515.0005BA
H-Glu(OME)-OtBu·HCl
H-Glu(OME)-OtBu·HCl
Sequence: H-Glu(OMe)-OtBu · HCl
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₀H₂₀ClNO₄ Storage Temperature: Freezer |
MDL Number:
MFCD02094380 CAS Number: 34582-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...