L-Cystine bisallyl ester di(p-toluenesulfonate) salt

Supplier: BACHEM AMERICAS INC

Synonyms: (H-Cys-allyl ester)₂·2 p-tosylate

4028317.0005 4028317.0025
E-3465.0005BAEA 361.41 CAD
E-3465.0005BA E-3465.0025BA
L-Cystine bisallyl ester di(p-toluenesulfonate) salt
L-Cystine bisallyl ester di(p-toluenesulfonate) salt

Sequence: (H-Cys-allyl ester)₂ · 2 p-tosylate


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.


Educt for Fmoc-Cys-OAll.

Formula: C₂₆H₃₆N₂O₁₀S₄
Storage Temperature: Freezer
MDL Number: MFCD00043424
CAS Number: 142601-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR