Dde-Lys(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, DDE-Lys(Fmoc)-OH

4027145.0001 4027145.0005
E-3385.0001BAEA 569.87 CAD
E-3385.0001BA E-3385.0005BA
Dde-Lys(Fmoc)-OH
Dde-Lys(Fmoc)-OH

Sequence: Dde-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


The amino-protecting group Dde is orthogonal to both Fmoc and Boc protection and finds increasing application in the synthesis of modified peptides assembled by Fmoc-tBu solid-phase methodologies. Dde can be cleaved by 2 % hydrazine in DMF.

Formula: C₃₁H₃₆N₂O₆
MW: 532.637 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467662
CAS Number: 156648-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR