H-Trp(Boc)-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Trp(Boc)-OtBu·HCl

4027105.0001 4027105.0005
E-3370.0001BAEA 884.75 CAD
E-3370.0001BA E-3370.0005BA
H-Trp(Boc)-OtBu·HCl
H-Trp(Boc)-OtBu·HCl

Sequence: H-Trp(Boc)-OtBu · HCl

Formula: C₂₀H₂₉ClN₂O₄
MW: 396.912 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798682
CAS Number: 218938-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR