H-Arg(Pbf)-OtBu · HCl

Supplier: BACHEM AMERICAS INC

4026726.0001 4026726.0005
E-3330.0001BAEA 828.77 CAD
E-3330.0001BA E-3330.0005BA
H-Arg(Pbf)-OtBu · HCl
H-Arg(Pbf)-OtBu · HCl

Sequence: H-Arg(Pbf)-OtBu · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₂₃H₃₈N₄O₅S·HCl
Storage Temperature: Freezer
MDL Number: MFCD08458588
CAS Number: 1217317-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR