H-Arg(Pbf)-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-Methyl 2-amino-5-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)guanidino)pentanoate hydrochloride, H-Arg(Pbf)-OMe·HCl, (S)-Methyl 2-amino-5-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulphonyl)guanidino)pentanoate hydrochloride

4026725.0001 4026725.0005
E-3325.0001BAEA 232.35 CAD
E-3325.0001BA E-3325.0005BA
H-Arg(Pbf)-OMe·HCl
H-Arg(Pbf)-OMe·HCl

Sequence: H-Arg(Pbf)-OMe · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₂₀H₃₃ClN₄O₅S
MW: 477.017 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318755
CAS Number: 257288-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR