H-Glu-otbu

Supplier: BACHEM AMERICAS INC

4030629.0001 4030629.0005
E-3295.0001BAEA 258.14 CAD
E-3295.0001BA E-3295.0005BA
H-Glu-otbu
H-Glu-otbu

Sequence: H-Glu-OtBu


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₉H₁₇NO₄
Storage Temperature: Freezer
MDL Number: MFCD00038562
CAS Number: 45120-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR