H-Gln(isopropyl)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-5-(isopropylamino)-5-oxopentanoic acid

4026437.0001 4026437.0005
E-3285.0001BAEA 309.81 CAD
E-3285.0001BA E-3285.0005BA
H-Gln(isopropyl)-OH
H-Gln(isopropyl)-OH

Sequence: H-Gln(isopropyl)-OH


For isoglutamine derivatives please see the family "Glutamic Acid".


An Nδ-alkylated Gln, which was incorporated in LHRH antagonists.

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672360
CAS Number: 4311-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR