O-Methyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

4003985.0001 4003985.0005 4003985.0025
E-3260.0001BAEA 358.5 CAD
E-3260.0001BA E-3260.0005BA E-3260.0025BA
O-Methyl-L-tyrosine
O-Methyl-L-tyrosine

Sequence: H-Tyr(Me)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
p-Methoxyphenylalanine and p-ethoxyphenylalanine.


Synonym(s): H-4-Methoxy-Phe-OH

Formula: C₁₀H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00002604
CAS Number: 6230-11-1
EINECS: 228-333-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR