L-Abrine

Supplier: BACHEM AMERICAS INC

Synonyms: Nalpha-Methyl-L-tryptophan

4026253.0250 4026253.1000
E-3255.0250BAEA 250.37 CAD
E-3255.0250BA E-3255.1000BA
L-Abrine
L-Abrine

Sequence: N-Me-Trp-OH
Synonym(s): L-Abrine


MeTrp, αMeTrp.

Formula: C₁₂H₁₄N₂O₂
MW: 218.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005645
CAS Number: 526-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR