N-Me-Asp(Otbu)-OH
Supplier: BACHEM AMERICAS INC
4012095.0250
4012095.1000
E-3190.0250BAEA
206.51
CAD
E-3190.0250BA
E-3190.1000BA
N-Me-Asp(Otbu)-OH
N-Me-Asp(Otbu)-OH
Sequence: N-Me-Asp(OtBu)-OH
Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Bachem also offers the neurotransmitter NMDA, F-2145.
Formula:
C₉H₁₇NO₄ Storage Temperature: Freezer |
MDL Number:
MFCD00672353 CAS Number: 197632-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...