N-Me-Lys(Z)-OH

Supplier: BACHEM AMERICAS INC

4025360.0250 4025360.1000
E-3185.0250BAEA 232.35 CAD
E-3185.0250BA E-3185.1000BA
N-Me-Lys(Z)-OH
N-Me-Lys(Z)-OH

Sequence: N-Me-Lys(Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₅H₂₂N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00672352
CAS Number: 201016-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR