N-Me-Lys-OH

Supplier: BACHEM AMERICAS INC

4016705.0250 4016705.1000
E-3180.0250BAEA 361.41 CAD
E-3180.0250BA E-3180.1000BA
N-Me-Lys-OH
N-Me-Lys-OH

Sequence: N-Me-Lys-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₇H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00672351
CAS Number: 7431-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR