N-(2-Aminoethyl)glycine

Supplier: BACHEM AMERICAS INC

4025636.0001 4025636.0005
E-3175.0001BAEA 232.35 CAD
E-3175.0001BA E-3175.0005BA
N-(2-Aminoethyl)glycine
N-(2-Aminoethyl)glycine

Sequence: N-β-Aminoethyl-Gly-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.


Aeg, educt for building blocks of PNA.

Formula: C₄H₁₀N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00144824
CAS Number: 24123-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR