H-Pro-NHEt·HCl
Supplier: BACHEM AMERICAS INC
4006174.0001
4006174.0005
E-3140.0001BAEA
309.66
CAD
E-3140.0001BA
E-3140.0005BA
H-Pro-NHEt·HCl
H-Pro-NHEt·HCl
Sequence: H-Pro-NHEt · HCl
The C-terminal residue of highly active GnRH agonists as buserelin or leuprorelin replacing the C-terminal Pro-Gly amide of the native peptide hormone.
Formula:
C₇H₁₅ClN₂O Storage Temperature: Freezer |
MDL Number:
MFCD00238159 CAS Number: 58107-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...