(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride
4019074.0001
4019074.0005
4019074.0025
E-3115.0001BAEA
185.46
CAD
E-3115.0001BA
E-3115.0005BA
E-3115.0025BA
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
Sequence: H-Asp(OMe)-OH · HCl
Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.
Formula:
C₅H₉NO₄·HCl MW: 183.6 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038972 CAS Number: 16856-13-6 EINECS: 240-880-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...