H-Arg(pbf)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: H-Arg(Pbf)-OH

4019730.0001 4019730.0005 4019730.0025
E-3110.0001BAEA 180.91 CAD
E-3110.0001BA E-3110.0005BA E-3110.0025BA
H-Arg(pbf)-OH
H-Arg(pbf)-OH

Sequence: H-Arg(Pbf)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₁₉H₃₀N₄O₅S
MW: 426.532 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236797
CAS Number: 200115-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR