(L)-Tryptophan-t-butyl ester
Supplier: BACHEM AMERICAS INC
4018557.0005
4018557.0025
4018557.0001
E-3105.0005BAEA
361.41
CAD
E-3105.0005BA
E-3105.0025BA
E-3105.0001BA
(L)-Tryptophan-t-butyl ester
(L)-Tryptophan-t-butyl ester
CAS: 16874-09-2 net
Formula: C15H20N2O2 · HCl
M.W: 296.8
Storage: -20 ± 5 °C
Formula:
C₁₅H₂₀N₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD00238204 CAS Number: 16874-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...