H-ARG(PMC)-OtBu

Supplier: BACHEM AMERICAS INC

4018591.0001 4018591.0005
E-3085.0001BAEA 828.77 CAD
E-3085.0001BA E-3085.0005BA
H-ARG(PMC)-OtBu
H-ARG(PMC)-OtBu

Sequence: H-Arg(Pmc)-OtBu (free base)


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.


Used in the synthesis of peptide primers for cyanophycin, a cyanobacterial reserve polymer.

Formula: C₂₄H₄₀N₄O₅S
Storage Temperature: Freezer
MDL Number: MFCD00236799
CAS Number: 169543-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR