L-Phenylalanine benzyl ester hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Phe-OBzl.HCl, H-Phe-OBzl·HCl
4017515.0005
4017515.0025
E-3080.0005BAEA
180.91
CAD
E-3080.0005BA
E-3080.0025BA
L-Phenylalanine benzyl ester hydrochloride
L-Phenylalanine benzyl ester hydrochloride
Sequence: H-Phe-OBzl · HCl
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Formula:
C₁₆H₁₇NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00043249 CAS Number: 2462-32-0 EINECS: 219-558-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...