H-Arg(Mtr)-OtBu (free base)

Supplier: BACHEM AMERICAS INC

4018553.0001 4018553.0005
E-3075.0001BAEA 593.81 CAD
E-3075.0001BA E-3075.0005BA
H-Arg(Mtr)-OtBu (free base)
H-Arg(Mtr)-OtBu (free base)

Sequence: H-Arg(Mtr)-OtBu (free base)


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₂₀H₃₄N₄O₅S
MW: 442.575 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236795
CAS Number: 115608-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR