α-Aminoisobutyric acid t-butyl ester

Supplier: BACHEM AMERICAS INC

4019068.0001 4019068.0005 4019068.0025
E-3060.0001BAEA 180.91 CAD
E-3060.0001BA E-3060.0005BA E-3060.0025BA
α-Aminoisobutyric acid t-butyl ester
alpha-Aminoisobutyric acid t-butyl ester

Sequence: H-Aib-OtBu · HCl
Synonym(s): H-α-Me-Ala-OtBu · HCl

Formula: C₈H₁₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08272284
CAS Number: 4512-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR