AC-Thr(tBu)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
(2S,3R)-2-Acetamido-3-(tert-butoxy)butanoic acid
4018967.0001
4018967.0005
E-3050.0001BAEA
361.41
CAD
E-3050.0001BA
E-3050.0005BA
AC-Thr(tBu)-OH
AC-Thr(tBu)-OH
Sequence: Ac-Thr(tBu)-OH
Bachem also offers phosphothreonine derivatives.
Formula:
C₁₀H₁₉NO₄ MW: 217.265 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00236771 CAS Number: 163277-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...