Ac-Glu(OtBu)-OH

Supplier: BACHEM AMERICAS INC

4018525.0001 4018525.0005
E-3040.0001BAEA 335.58 CAD
E-3040.0001BA E-3040.0005BA
Ac-Glu(OtBu)-OH
Ac-Glu(OtBu)-OH

Sequence: Ac-Glu(OtBu)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₁H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00236754
CAS Number: 84192-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR