AC-Ser(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Acetamido-3-(tert-butoxy)propanoic acid, N-Acetyl-O-tert-butyl-L-serine

4017866.0001 4017866.0005
E-3010.0001BAEA 361.41 CAD
E-3010.0001BA E-3010.0005BA
AC-Ser(tBu)-OH
AC-Ser(tBu)-OH

Sequence: Ac-Ser(tBu)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236770
CAS Number: 77285-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR